Stampa…
Water stills for single and double distillation
Fornitore: GFL
171-3042EA 5850 EUR
GFLA2302 GFLA2202 GFLA2304 GFLA2204 GFLA2208
Water stills for single and double distillation
These water purification systems produce low-gas, bacteria and pyrogen free distillate with a very low conductivity.
  • Fully automatic
  • Bench and wall mounting systems
  • Quartz sleeved heating element
  • Condensers are kept sterile by steam
  • Cooling water outlet with 12,7 mm Int.Ø tubing connection
Evaporators, condensers and overflows made of DURAN®/Borosilicate glass 3.3 and housings are made of electrolytically galvanised sheet steel, electrostatically powder-coated with epoxy resin.
After the main switch is switched on, the solenoid valve opens to start the water supply. Water will not be condensed flows out through the cooling water outlet. Low energy consumption through distilling the heated cooling water. Distillate withdrawal at the right-hand side of the unit through tubing connection. Electronic level control during the whole distillation process. Water level control in the condenser has automatic power cut-off in the event of water shortage. Electronic impurity detector starts automatic water change to rinse and clean the condenser. Degassing of carbon dioxide through vents on the condenser. Main and function switches as well as pilot lamps for monitoring are on the front of the unit. Visible distillation process through easily removable, unbreakable front screen that does not steam up.
Power connection through connection cable models 2202, 2204 and 2302 with German shock-proof type (Schuko) plug.
Certificazioni: Conformity with DAB requirements and international pharmacopoeia regulations.