prepGEM® DNA extraction kits have been developed to provide a rapid, reliable, and economical solution to DNA extraction. The kits use a single tube protocol with temperature control to prepare DNA for analysis, and are optimised for downstream PCR-based applications, including qPCR, STR-PCR, DNA sequencing, AFLP analysis, and many SNiP detection applications.
prepGEM® Blood kit is a simple enzymatic DNA extraction system specifically optimised to overcome the PCR inhibition commonly found with single tube extraction protocols for blood. It is ideal for low to high throughput biomedical research applications, which use PCR, qPCR, STR-PCR, or WGA based genotyping protocols.
prepGEM® Blood kit is a simple enzymatic DNA extraction system specifically optimised to overcome the PCR inhibition commonly found with single tube extraction protocols for blood. It is ideal for low to high throughput biomedical research applications, which use PCR, qPCR, STR-PCR, or WGA based genotyping protocols.