Italy
Order Entry
Nitric acid fuming 90+% ACS
 
 
 
Nitric acid fuming 90+% ACS
  ACRO270620010
 :  
undefined
 
 
 
Nitric acid fuming 90+% ACS
  ACRO270620010
 :  
 : 270620010
 : ACRO270620010
 
270,00 €
   
  1 * 1 L

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Residue after ignition:
    ≤0.002 %
  • Titration with NaOH:
    ≥90.0 %
  • Dilution test:
    Passes test
  • Chloride (Cl):
    ≤0.7 ppm
  • Sulfate (SO4):
    ≤5 ppm
  • Arsenic (As):
    ≤0.3 ppm
  • Iron (Fe):
    ≤2 ppm
  • Heavy metals (as Pb):
    ≤5 ppm
  • Dissolved oxides:
    ≤0.1 % (as N2O3)

 

 

 :
 :63.01 g/mol
 :13,06610
 :84 °C (760 mmHg)
 :–40 °C
 :1.511 g/cm³
 :Ambient
 :231-714-2
 :2032
 :8,I
 :01-2119487297-23
 :GHS03
 :GHS05
 :GHS06
 :7697-37-2
 :HNO₃
 :MFCD00011349