Shakers, rocker 2D and 3D basic and digital
: IKA
- Catalog No:
- 444-0817
- 444-0816
- 444-0815
- 444-0818
- Model:
- Rocker 3D basic
- Rocker 2D digital
- Rocker 2D basic
- Rocker 3D digital
- Shaking motion:
- Tumbling
- Rocking
- Rocking
- Tumbling
- Speed range:
- 30 min⁻¹ (fixed)
- 5 - 80 min⁻¹
- 5 - 80 min⁻¹
- 5 - 80 min⁻¹
- Angle:
- 8°
- 0 - 15°
- 8°
- 0 - 15°
- W×D×H:
- 280×330×165 mm
- 280×330×150 mm
- 280×330×150 mm
- 280×330×185 mm
- Weight:
- 2,2 kg
- 2,2 kg
- 2,2 kg
- 2,2 kg
- Max. load:
- 2 kg
- 2 kg
- 2 kg
- 2 kg
- Pk
- Rocker 2D basicRocker 2D basic444-0815400200011 * 1 pezzi1 069,00 €
Accessories
Specifications
ModelRocker 2D basicEnvironmentally PreferableShaking motionRockingSpeed range5 - 80 min⁻¹Angle8°W×D×H280×330×150 mmWeight2,2 kgMax. load2 kg - Rocker 2D digitalRocker 2D digital444-0816400300011 * 1 pezzi1 137,00 €
Accessories
Specifications
ModelRocker 2D digitalEnvironmentally PreferableShaking motionRockingSpeed range5 - 80 min⁻¹Angle0 - 15°W×D×H280×330×150 mmWeight2,2 kgMax. load2 kg - Rocker 3D basicRocker 3D basic444-0817400000011 * 1 pezzi1 137,00 €
Accessories
Specifications
ModelRocker 3D basicEnvironmentally PreferableShaking motionTumblingSpeed range30 min⁻¹ (fixed)Angle8°W×D×H280×330×165 mmWeight2,2 kgMax. load2 kg - Rocker 3D digitalRocker 3D digital444-0818400100011 * 1 pezzi1 201,00 €
Accessories
Specifications
ModelRocker 3D digitalEnvironmentally PreferableShaking motionTumblingSpeed range5 - 80 min⁻¹Angle0 - 15°W×D×H280×330×185 mmWeight2,2 kgMax. load2 kg