Tris(pentafluorophenyl)borane ≥97%
Fornitore: Thermo Scientific
|
Pericolo
|
L18054.03EA
172
EUR
L18054.03
L18054.06
Tris(pentafluorophenyl)borane ≥97%
Tris(pentafluorophenyl)borane
Formula:
C₁₈BF₁₅ Peso molecolare: 511,98 g/mol Punto di fusione: 126…130 °C |
MDL Number:
MFCD00269813 CAS Number: 1109-15-5 EINECS: 213-504-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...