Stampa…
Toluene[methyl-¹³C] (99% ¹³C )
Fornitore: LGC Standards PROMOCHEM
Pericolo
PROOCILCLM-309-0.5EA 818 EUR
PROOCILCLM-309-0.5 PROOCIL-CLM-309-1
Toluene[methyl-¹³C] (99% ¹³C )
Toluene[methyl-¹³C]
Formula: C₆H₅CH₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00084093
CAS Number: 6933-23-9
UN: 1294
ADR: 3,II