Safety padlocks, aluminium
Supplier: Brady
Certificates
About this item
Manufactured with solid aluminium body and shackles, these padlocks are amazingly lightweight and rugged.
- Locks are chemical and corrosion resistant and feature 6-pin cylinders for better tamper resistance
- Padlocks are supplied keyed different
- Two keys supplied with each lock, NOT key retaining
Shackle clearance: 20,00 mm
Shackle width: 31,50 mm
Shackle height 25 mm
Product Information
- Catalog #Supplier #TypeColourW×D×HPkAvailabilityPrice
- BRDY814071814071Shackle height 25 mmLilac40×19×73 mm61 * 6 pezzi179,66 €Supplier Article Number: 814071Local Article Number: BRDY814071
Specifications
Environmentally Preferable - BRDY814066814066Shackle height 25 mmRed40×19×73 mm61 * 6 pezzi179,66 €Supplier Article Number: 814066Local Article Number: BRDY814066
Specifications
Environmentally Preferable - BRDY814067814067Shackle height 25 mmBlack40×19×73 mm61 * 6 pezzi179,66 €Supplier Article Number: 814067Local Article Number: BRDY814067
Specifications
Environmentally Preferable - BRDY814068814068Shackle height 25 mmYellow40×19×73 mm61 * 6 pezzi179,66 €Supplier Article Number: 814068Local Article Number: BRDY814068
Specifications
Environmentally Preferable - BRDY814070814070Shackle height 25 mmOrange40×19×73 mm61 * 6 pezzi179,66 €Supplier Article Number: 814070Local Article Number: BRDY814070
Specifications
Environmentally Preferable - BRDY830939830939Shackle height 25 mmBrown40×19×73 mm61 * 6 pezzi179,66 €Supplier Article Number: 830939Local Article Number: BRDY830939
Specifications
Environmentally Preferable - BRDY814065814065Shackle height 25 mmBlue40×19×73 mm61 * 6 pezzi179,66 €Supplier Article Number: 814065Local Article Number: BRDY814065
Specifications
Environmentally Preferable - BRDY830936830936Shackle height 25 mmGrey40×19×73 mm61 * 6 pezzi179,66 €Supplier Article Number: 830936Local Article Number: BRDY830936
Specifications
Environmentally Preferable - BRDY814069814069Shackle height 25 mmGreen40×19×73 mm61 * 6 pezzi179,66 €Supplier Article Number: 814069Local Article Number: BRDY814069
Specifications
Environmentally Preferable
Shackle height 40 mm
Product Information
- Catalog #Supplier #TypeColourW×D×HPkAvailabilityPrice
- BRDY814074814074Shackle height 40 mmBlack40×19×89 mm61 * 6 pezzi184,00 €Supplier Article Number: 814074Local Article Number: BRDY814074
Specifications
Environmentally Preferable - BRDY814072814072Shackle height 40 mmBlue40×19×89 mm61 * 6 pezzi184,00 €Supplier Article Number: 814072Local Article Number: BRDY814072
Specifications
Environmentally Preferable - BRDY814073814073Shackle height 40 mmRed40×19×89 mm61 * 6 pezzi184,00 €Supplier Article Number: 814073Local Article Number: BRDY814073
Specifications
Environmentally Preferable - BRDY814076814076Shackle height 40 mmGreen40×19×89 mm61 * 6 pezzi184,00 €Supplier Article Number: 814076Local Article Number: BRDY814076
Specifications
Environmentally Preferable - BRDY814075814075Shackle height 40 mmYellow40×19×89 mm61 * 6 pezzi184,00 €Supplier Article Number: 814075Local Article Number: BRDY814075
Specifications
Environmentally Preferable - BRDY830937830937Shackle height 40 mmGrey40×19×89 mm61 * 6 pezzi184,00 €Supplier Article Number: 830937Local Article Number: BRDY830937
Specifications
Environmentally Preferable - BRDY814078814078Shackle height 40 mmLilac40×19×89 mm61 * 6 pezzi184,00 €Supplier Article Number: 814078Local Article Number: BRDY814078
Specifications
Environmentally Preferable - BRDY814077814077Shackle height 40 mmOrange40×19×89 mm61 * 6 pezzi184,00 €Supplier Article Number: 814077Local Article Number: BRDY814077
Specifications
Environmentally Preferable - BRDY830940830940Shackle height 40 mmBrown40×19×89 mm61 * 6 pezzi184,00 €Supplier Article Number: 830940Local Article Number: BRDY830940
Specifications
Environmentally Preferable
Shackle height 75 mm
Product Information
- Catalog #Supplier #TypeColourW×D×HPkAvailabilityPrice
- BRDY814079814079Shackle height 75 mmBlue40×19×119 mm61 * 6 pezzi192,67 €Supplier Article Number: 814079Local Article Number: BRDY814079
Specifications
Environmentally Preferable - BRDY814081814081Shackle height 75 mmBlack40×19×119 mm61 * 6 pezzi192,67 €Supplier Article Number: 814081Local Article Number: BRDY814081
Specifications
Environmentally Preferable - BRDY814082814082Shackle height 75 mmYellow40×19×119 mm61 * 6 pezzi192,67 €Supplier Article Number: 814082Local Article Number: BRDY814082
Specifications
Environmentally Preferable - BRDY814083814083Shackle height 75 mmGreen40×19×119 mm61 * 6 pezzi192,67 €Supplier Article Number: 814083Local Article Number: BRDY814083
Specifications
Environmentally Preferable - BRDY814085814085Shackle height 75 mmLilac40×19×119 mm61 * 6 pezzi192,67 €Supplier Article Number: 814085Local Article Number: BRDY814085
Specifications
Environmentally Preferable - BRDY814084814084Shackle height 75 mmOrange40×19×119 mm61 * 6 pezzi192,67 €Supplier Article Number: 814084Local Article Number: BRDY814084
Specifications
Environmentally Preferable - BRDY830938830938Shackle height 75 mmGrey40×19×119 mm61 * 6 pezzi192,67 €Supplier Article Number: 830938Local Article Number: BRDY830938
Specifications
Environmentally Preferable - BRDY814080814080Shackle height 75 mmRed40×19×119 mm61 * 6 pezzi192,67 €Supplier Article Number: 814080Local Article Number: BRDY814080
Specifications
Environmentally Preferable - BRDY830941830941Shackle height 75 mmBrown40×19×119 mm61 * 6 pezzi192,67 €Supplier Article Number: 830941Local Article Number: BRDY830941