Stampa…

Triphenylantimony dichloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

TCIAT1849-1GEA 66 EUR
TCIAT1849-1G
Triphenylantimony dichloride ≥98.0% (by titrimetric analysis)
Triphenylantimony dichloride
Formula: C₁₈H₁₅Cl₂Sb
Peso molecolare: 423,98 g/mol
MDL Number: MFCD00013598
CAS Number: 594-31-0
EINECS: 209-834-1
UN: 3467
ADR: 6.1,III
https://it.vwr.com/stibo/web/std.lang.all/58/20/28995820.jpg

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 142.0 to 145.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR