Furfuryl isopropyl sulphide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
TCIAF0742-25GEA
239
EUR
TCIAF0742-25G
Furfuryl isopropyl sulphide ≥98.0% (by GC)
Furfuryl isopropyl sulphide
Formula:
C₈H₁₂OS Peso molecolare: 156,25 g/mol Punto di ebollizione: 80°C Flash Pt: 76°C |
MDL Number:
MFCD00040265 CAS Number: 1883-78-9 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...