Stampa…
Hai cercato: Tester di rivestimento
46  risultati sono stati trovati
SearchResultCount:"46"
Sort Results
Visualizzazione a lista Nuova Visualizzazione
La tua valutazione
Fornitore: Merck
Descrizione: Colorstream® multicolor effect pigments add dimensions to styling possibilities: They can change colour with the angle of observation, allowing innova...
Fornitore: Industrial Physics
Descrizione: The Colorbox is used to provide the illumination conditions for any visual inspection at a 45° angle.
Codice catalogo: (SIMESP1896)
Fornitore: Industrial Physics
Codice articolo fornitore: SP1896
Codice articolo locale: SIMESP1896
Descrizione: Impact test used to determine the impact resistivity and flexibility of materials.
UOM: 1 * 1 pezzi
Fornitore: Industrial Physics
Descrizione: These cylindrical bend testers are used to determine the elasticity, adhesion and elongation of paint on metal.
Fornitore: Industrial Physics
Descrizione: This coating thickness gauge is a fully electronic apparatus, that uses magnetic and eddy current principles, to measure coating thickness on both fer...
Fornitore: Industrial Physics
Descrizione: The Positector 200 has ultrasound technology to measure coating thickness over non-metal substrates.
Fornitore: Industrial Physics
Descrizione: Calibration standards are meant for verifying coating gauges - These plastic foils and shims are acting as standards.
Fornitore: Industrial Physics
Descrizione: This stiffness tester is used to assess material stiffness and material resiliency for materials such as paper products, cardboard, plastics, textiles...
Fornitore: Industrial Physics
Descrizione: Electronic coating gauges use magnetic and eddy current principles to measure coating thickness on both ferrous and non-ferrous metals with all PosiTe...
Fornitore: Industrial Physics
Descrizione: Test panels are used to enhance reproducibility of physical and chemical tests. Each panel is equipped with a hole for hanging and handling.
Fornitore: Industrial Physics
Descrizione: These digital spring micrometer measures and records peak to valley surface profile height using replica tape reader and it provides simple, durable, ...
Fornitore: Industrial Physics
Descrizione: This apparatus is designed to detect of coating porosity in dielectric (insulation type) coatings on conductive substrates, including concrete.
Fornitore: Industrial Physics
Descrizione: These gloss meters are designed for the measurement of gloss levels on flat surfaces.
Fornitore: Industrial Physics
Descrizione: These roughness scales are designed to estimate the surface roughness, at touch or sight, specifically developed for the profiles related to the condi...
Fornitore: Industrial Physics
Descrizione: The PosiTector can measure the wall thickness of materials (plastic, stell) using ultrasonic technology. These durable and weatherproof models are res...
Fornitore: Industrial Physics
Descrizione: This replica tape is specifically designed to measure blasted surfaces. By placing the Tqc replica tape on the surface and rubbing over it, the Rt (to...
Prezzo a richiesta
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.
Il prodotto originale non è più disponibile. L'articolo suggerito è disponibile
Il prodotto marcato con questo simbolo non è più disponibile o vendibile fino ad esaurimento scorte. Alternative possono essere disponibili utilizzando il numero di catalogo VWR. Per maggiori informazioni può contattare il nostro customer service al 023320311 o inviando una email all'indirizzo ebusiness.it@vwr.com
1 - 16 of 46
no targeter for Bottom