Stampa…
Blood sedimentation apparatus
Fornitore: Hecht
HECH40367010EA 168 EUR
HECH40367010
Blood sedimentation apparatus
Sistemi di diagnostica Strumenti clinici
Blood sedimentation apparatus stand of stainless steel with tilting device for rapid sedimentation.
  • Pipettes with fused graduation 'ASSISTENT-PERMANENT'
Informazioni per l'ordine: Complete with blood sedimentation pipettes, blood mixing glasses with rubber stopper, special rubber caps (macro method only) and directions for use.
Confezione: In a box.