Specifiche
Punto di fusione | 189-191° | |
Peso formula | 220.12 | |
CAS No. | 17518-49-9 | |
Purezza | 97% | |
Formula molecolare | C8H7Cl2NS | |
N. MDL | MFCD00052483 |
![]() |
Stiamo processando la tua richiesta… |
Articoli aggiunti al tuo carrello
|
||||||||
---|---|---|---|---|---|---|---|---|
|
Avantor Services provides a wide range of specialized services and digital solutions to help you solve complex challenges.
We’ve built our reputation on consistent, comprehensive mastery of day-to-day operations, allowing lab, clinical, and production environments to focus their high-value resources on core scientific priorities.
As our customers’ needs have evolved, so have our capabilities. We have become experts in scientific operations, improving performance with sophisticated solutions and providing guidance on best practices.
You can select and customize services for peak efficiency, quality, and accelerated innovation.
Contact us at services@avantorsciences.com.
![]() |
|
|
Avantor is a leading global provider of integrated, tailored solutions for the life sciences and advanced technology industries. Strengthened by the recent acquisition of VWR, the Company is a trusted partner to customers and suppliers from discovery to delivery. Collectively, we set science in motion to create a better world.
VWR International Srl
Via San Giusto 85-20153 Milano
Cod. Fisc. e Part IVA IT 12864800151 Codice identificativo CBBV91S