Stampa…
Di-n-octyl phthalate, Supelco®
Fornitore: Merck
M-Clarity™ quality level = MQ100
31301-250MGEA 40 EUR
31301-250MG
Di-n-octyl phthalate, Supelco®
Di-n-octyl phthalate
Formula: C₂₄H₃₈O₄
Peso molecolare: 390,56 g/mol
MDL Number: MFCD00015292
CAS Number: 117-84-0
EINECS: 204-214-7