5,6-Dibromo-2,1,3-benzothiadiazole ≥98.0% (by GC)
Fornitore: TCI EUROPE
TCIAD4064-5GEA
160
EUR
TCIAD4064-5G
5,6-Dibromo-2,1,3-benzothiadiazole ≥98.0% (by GC)
5,6-Dibromo-2,1,3-benzothiadiazole
Formula:
C₆H₂Br₂N₂S Peso molecolare: 293,9700 g/mol |
MDL Number:
MFCD20257910 CAS Number: 18392-81-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 131.0 to 135.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...