3,4,5,6-Tetrafluorosalicylic acid ≥98.0%
Fornitore: TCI EUROPE
|
Attenzione
|
TCIAT1988-1GEA
202
EUR
TCIAT1988-1G
3,4,5,6-Tetrafluorosalicylic acid ≥98.0%
3,4,5,6-Tetrafluorosalicylic acid
Formula:
C₇H₂F₄O₃ |
MDL Number:
MFCD00483242 CAS Number: 14742-36-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 170.0 to 175.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...