4,5-Dimethoxy-2-nitrobenzonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
TCIAD2984-5GEA
62
EUR
TCIAD2984-5G
4,5-Dimethoxy-2-nitrobenzonitrile ≥98.0% (by GC)
4,5-Dimethoxy-2-nitrobenzonitrile
Formula:
C₉H₈N₂O₄ |
MDL Number:
MFCD00017564 CAS Number: 102714-71-6 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 165.0 to 170.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...