3-Amino-3-(4-fluorophenyl)propionic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
TCIAA2430-1GEA
188
EUR
TCIAA2430-1G
3-Amino-3-(4-fluorophenyl)propionic acid ≥98.0% (by HPLC, titration analysis)
3-Amino-3-(4-fluorophenyl)propionic acid
Formula:
C₉H₁₀FNO₂ Peso molecolare: 183,18 g/mol Punto di fusione: 216°C |
MDL Number:
MFCD00181811 CAS Number: 325-89-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...