Stampa…
N-Boc-N-propylethylenediamine 96%
Fornitore: Thermo Scientific
Attenzione
H52328.MDEA 202 EUR
H52328.MD H52328.03
N-Boc-N-propylethylenediamine 96%
N-Boc-N-propylethylenediamine
Formula: C₁₀H₂₂N₂O₂
Peso molecolare: 202,29 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06658368
CAS Number: 105628-64-6