Stampa…

1-Amino-4-hydroxy-9,10-anthraquinone ≥98.0% (by total nitrogen basis)

Fornitore: TCI EUROPE

TCIAA0314-5GEA 266 EUR
TCIAA0314-5G
1-Amino-4-hydroxy-9,10-anthraquinone ≥98.0% (by total nitrogen basis)
1-Amino-4-hydroxy-9,10-anthraquinone
Formula: C₁₄H₉NO₃
Peso molecolare: 239,23 g/mol
MDL Number: MFCD00001223
CAS Number: 116-85-8
EINECS: 204-163-0
https://it.vwr.com/stibo/web/std.lang.all/77/49/28927749.jpg

Visualizza i codici

Risultati del test di specificazione

Appearance Green to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 212.0 to 218.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR