Stampa…
Isosorbide
Fornitore: LGC Standards PROMOCHEM
PROOMM0126.03EA 984 EUR
PROOMM0126.03
Isosorbide
Isosorbide
Formula: C₆H₁₀O₄
Peso molecolare: 146,14 g/mol
MDL Number: MFCD00064827
CAS Number: 652-67-5
EINECS: 211-492-3