Stampa…
Pipette holder
Fornitore: Karl Hecht
HECH42583010EA 39 EUR
HECH42583010
Pipette holder
This pipette holder for 12 pipettes is made of plexiglas®.
  • With numbered positions and field for inscriptions